Assay Design
Gene Transcripts
- Gene Symbol
- ALOX12P2
- Entrez Gene ID
-
245
- Gene Name
- arachidonate 12-lipoxygenase pseudogene 2
- Gene Aliases
-
ALOX12E
- Chromosome Location
-
Chr.17: 6853576 - 6900349 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002710.2 |
|
11 - 12 |
2020 |
78 |
NR_120453.1 |
|
10 - 11 |
1467 |
78 |
GenBank mRNA |
AL832768.1 |
- |
11 - 12 |
1737 |
78 |
BC041851.1 |
- |
13 - 14 |
2201 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA411551.1 |
- |
1 - 2 |
64 |
|
Chr. 17 - 6898868 - 6900094 [+] on Build GRCh38 |
78 |
GenBankmRNA |
BX110191.1 |
- |
2 - 3 |
280 |
|
Chr. 17 - 6898553 - 6900001 [+] on Build GRCh38 |
78 |
GenBankmRNA |
AA411550.1 |
- |
2 - 3 |
282 |
|
Chr. 17 - 6898548 - 6899897 [+] on Build GRCh38 |
78 |
GenBankmRNA |
DN831733.1 |
- |
3 - 4 |
340 |
|
Chr. 17 - 6898253 - 6899746 [+] on Build GRCh38 |
78 |
GenBankmRNA |
DN831732.1 |
- |
2 - 3 |
|
|
Chr. 17 - 6898215 - 6899705 [-] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tektin 1 |
TEKT1 |
|
-67.1kb
|
Anti-sense |
Chr. - 6799981 - 6831741 on Build GRCh38 |
Downstream |
ALOX12 antisense RNA 1 |
ALOX12-AS1 |
|
85.6kb
|
Anti-sense |
Chr. - 6985123 - 7012334 on Build GRCh38 |
View Full Product Details