Assay Design
Gene Transcripts
- Gene Symbol
- ALOX12P2
- Entrez Gene ID
-
245
- Gene Name
- arachidonate 12-lipoxygenase pseudogene 2
- Gene Aliases
-
ALOX12E
- Chromosome Location
-
Chr.17: 6853576 - 6900349 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120453.1 |
|
1 - 2 |
142 |
104 |
GenBank mRNA |
DB019000.1 |
- |
1 - 2 |
142 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tektin 1 |
TEKT1 |
|
-62.0kb
|
Anti-sense |
Chr. - 6799981 - 6831741 on Build GRCh38 |
Downstream |
ALOX12 antisense RNA 1 |
ALOX12-AS1 |
|
90.6kb
|
Anti-sense |
Chr. - 6985123 - 7012334 on Build GRCh38 |
View Full Product Details