Assay Design
Gene Transcripts
- Gene Symbol
- SSPN
- Entrez Gene ID
-
8082
- Gene Name
- sarcospan
- Gene Aliases
-
DAGA5, KRAG, NSPN, SPN1, SPN2
- Chromosome Location
-
Chr.12: 26122024 - 26234777 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC041929.1 |
- |
1 - 2 |
60 |
89 |
View Full Product Details