Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024041.1 | 2 - 3 | 181 | 156 | |
GenBank mRNA | AV733354.1 | - | 2 - 3 | 181 | 156 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | IL21 antisense RNA 1 | IL21-AS1 | -41.1kb | Anti-sense | Chr. - 122618983 - 122689156 on Build GRCh38 | |
Downstream | fibroblast growth factor 2 | FGF2 | BFGF, FGF-2, FGFB, HBGF-2 | 94.5kb | Anti-sense | Chr. - 122826708 - 122898235 on Build GRCh38 |