Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017020288.1 | XP_016875777.1 | 10 - 11 | 1203 | 109 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017020287.1 | XP_016875776.1 | 9 - 10 | 999 | 109 |