Assay Design



Gene Transcripts

Gene Symbol
DRGX
Entrez Gene ID
644168
Gene Name
dorsal root ganglia homeobox
Gene Aliases
DRG11, PRRXL1
Chromosome Location
Chr.10: 49364085 - 49396016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276451.1 NP_001263380.1 3 - 4 294 74
XM_011540089.2 XP_011538391.1 2 - 3 212 74
GenBank mRNA HY131276.1 - 3 - 4 294 74

View Full Product Details