Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013660.2 | NP_001013682.2 | 12 - 13 | 1929 | 83 |
XM_005270861.3 | XP_005270918.1 | 12 - 13 | 1456 | 83 | |
XM_011541451.2 | XP_011539753.1 | 12 - 13 | 1929 | 83 | |
XM_011541452.1 | XP_011539754.1 | 12 - 13 | 1456 | 83 | |
XM_017001271.1 | XP_016856760.1 | 10 - 11 | 1340 | 83 | |
GenBank mRNA | AK131302.1 | - | 12 - 13 | 1929 | 83 |