Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005270861.3 | XP_005270918.1 | 16 - 17 | 1809 | 115 |
XM_011541451.2 | XP_011539753.1 | 16 - 17 | 2282 | 115 | |
XM_017001271.1 | XP_016856760.1 | 14 - 15 | 1693 | 115 |