Assay Design



Gene Transcripts

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
Chromosome Location
Chr.15: 34343315 - 34357737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284293.1 NP_001271222.1 1 - 2 390 72
GenBank mRNA AK302680.1 - 1 - 2 127 72

View Full Product Details