Assay Design



Gene Transcripts

Gene Symbol
PARP14
Entrez Gene ID
54625
Gene Name
poly(ADP-ribose) polymerase family member 14
Gene Aliases
ARTD8, BAL2, PARP-14, pART8
Chromosome Location
Chr.3: 122680726 - 122730840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017554.2 NP_060024.2 2 - 3 381 125
XM_011512928.2 XP_011511230.1 2 - 3 472 125
XM_011512929.2 XP_011511231.1 2 - 3 480 125
GenBank mRNA AK304269.1 - 2 - 3 349 125
CR977108.1 - 2 - 3 341 125
DC320673.1 - 2 - 3 383 125

View Full Product Details