Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213600.3 | NP_998765.3 | 2 - 3 | 279 | 69 |
GenBank mRNA | AK122623.1 | - | 2 - 3 | 250 | 69 |
AK172836.1 | - | 2 - 3 | 279 | 69 | |
BC131531.1 | - | 2 - 3 | 256 | 69 | |
BC142682.1 | - | 2 - 3 | 250 | 69 | |
BC146648.1 | - | 2 - 3 | 250 | 69 |