Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213600.3 | NP_998765.3 | 6 - 7 | 627 | 65 |
NR_033151.1 | 5 - 6 | 554 | 65 | ||
GenBank mRNA | AK122623.1 | - | 6 - 7 | 598 | 65 |
AK172836.1 | - | 6 - 7 | 627 | 65 | |
AK304368.1 | - | 6 - 7 | 609 | 65 | |
BC035335.1 | - | 5 - 6 | 524 | 65 | |
BC131531.1 | - | 6 - 7 | 604 | 65 | |
BC142682.1 | - | 6 - 7 | 598 | 65 | |
BC146648.1 | - | 6 - 7 | 598 | 65 |