Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 10 - 11 | 1333 | 71 |
XM_011540849.1 | XP_011539151.1 | 10 - 11 | 1416 | 71 | |
XM_011540850.2 | XP_011539152.1 | 10 - 11 | 1345 | 71 | |
XM_011540851.1 | XP_011539153.1 | 10 - 11 | 1300 | 71 | |
XM_011540852.2 | XP_011539154.1 | 10 - 11 | 1282 | 71 | |
XM_011540855.1 | XP_011539157.1 | 8 - 9 | 1138 | 71 | |
XM_011540857.1 | XP_011539159.1 | 8 - 9 | 897 | 71 | |
XM_011540859.2 | XP_011539161.1 | 7 - 8 | 866 | 71 | |
XM_017000490.1 | XP_016855979.1 | 10 - 11 | 1277 | 71 | |
XM_017000491.1 | XP_016855980.1 | 10 - 11 | 1277 | 71 | |
XM_017000492.1 | XP_016855981.1 | 4 - 5 | 380 | 71 | |
XM_017000494.1 | XP_016855983.1 | 10 - 11 | 1277 | 71 | |
XM_017000495.1 | XP_016855984.1 | 10 - 11 | 1277 | 71 | |
XM_017000496.1 | XP_016855985.1 | 10 - 11 | 1277 | 71 | |
GenBank mRNA | AK096330.1 | - | 9 - 10 | 1277 | 71 |
AK294754.1 | - | 4 - 5 | 350 | 71 | |
AK302582.1 | - | 6 - 7 | 501 | 71 | |
BC144198.1 | - | 9 - 10 | 1272 | 71 | |
DT932601.1 | - | 2 - 3 | 163 | 71 |