Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 13 - 14 | 1790 | 150 |
XM_011540849.1 | XP_011539151.1 | 13 - 14 | 1873 | 150 | |
XM_011540850.2 | XP_011539152.1 | 13 - 14 | 1802 | 150 | |
XM_011540851.1 | XP_011539153.1 | 13 - 14 | 1757 | 150 | |
XM_011540852.2 | XP_011539154.1 | 13 - 14 | 1739 | 150 | |
XM_011540855.1 | XP_011539157.1 | 11 - 12 | 1595 | 150 | |
XM_011540857.1 | XP_011539159.1 | 11 - 12 | 1354 | 150 | |
XM_011540859.2 | XP_011539161.1 | 10 - 11 | 1323 | 150 | |
XM_017000490.1 | XP_016855979.1 | 13 - 14 | 1734 | 150 | |
XM_017000491.1 | XP_016855980.1 | 13 - 14 | 1734 | 150 | |
XM_017000492.1 | XP_016855981.1 | 7 - 8 | 837 | 150 | |
XM_017000494.1 | XP_016855983.1 | 13 - 14 | 1734 | 150 | |
XM_017000495.1 | XP_016855984.1 | 13 - 14 | 1734 | 150 | |
XM_017000496.1 | XP_016855985.1 | 13 - 14 | 1734 | 150 | |
GenBank mRNA | AB058636.1 | - | 13 - 14 | 1791 | 150 |
AB058637.1 | - | 12 - 13 | 1733 | 150 | |
AB204867.1 | - | 13 - 14 | 1791 | 150 | |
AK294754.1 | - | 7 - 8 | 807 | 150 | |
AK302582.1 | - | 9 - 10 | 958 | 150 |