Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017554.2 | NP_060024.2 | 4 - 5 | 647 | 150 |
XM_011512928.2 | XP_011511230.1 | 4 - 5 | 738 | 150 | |
XM_011512929.2 | XP_011511231.1 | 4 - 5 | 746 | 150 | |
GenBank mRNA | AK304269.1 | - | 4 - 5 | 615 | 150 |
BU170133.1 | - | 1 - 2 | 161 | 150 | |
CR977108.1 | - | 4 - 5 | 607 | 150 | |
DQ063584.1 | - | 1 - 2 | 111 | 150 | |
DQ063585.1 | - | 1 - 2 | 111 | 150 |