Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018039.1 | NP_001018049.1 | 16 - 17 | 1905 | 64 |
NM_001029880.2 | NP_001025051.1 | 16 - 17 | 2007 | 64 | |
XM_005252551.2 | XP_005252608.1 | 16 - 17 | 1788 | 64 | |
XM_006717490.2 | XP_006717553.1 | 14 - 15 | 1780 | 64 | |
XM_011519611.1 | XP_011517913.1 | 16 - 17 | 2006 | 64 | |
XM_017016466.1 | XP_016871955.1 | 16 - 17 | 1908 | 64 | |
GenBank mRNA | AB046837.1 | - | 16 - 17 | 1905 | 64 |
BC152430.1 | - | 16 - 17 | 1905 | 64 |