Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018039.1 | NP_001018049.1 | 19 - 20 | 2504 | 58 |
NM_001029880.2 | NP_001025051.1 | 19 - 20 | 2606 | 58 | |
XM_005252551.2 | XP_005252608.1 | 19 - 20 | 2387 | 58 | |
XM_006717490.2 | XP_006717553.1 | 17 - 18 | 2379 | 58 | |
XM_011519611.1 | XP_011517913.1 | 19 - 20 | 2605 | 58 | |
XM_017016466.1 | XP_016871955.1 | 19 - 20 | 2507 | 58 | |
GenBank mRNA | AB046837.1 | - | 19 - 20 | 2504 | 58 |
BC144327.1 | - | 14 - 15 | 2074 | 58 | |
BC152430.1 | - | 19 - 20 | 2504 | 58 |