Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167962.1 | NP_001161434.1 | 4 - 5 | 548 | 71 |
NM_001321787.1 | NP_001308716.1 | 4 - 5 | 548 | 71 | |
NM_198999.2 | NP_945350.1 | 4 - 5 | 548 | 71 | |
NM_206883.2 | NP_996766.1 | 4 - 5 | 548 | 71 | |
NM_206884.2 | NP_996767.1 | 4 - 5 | 548 | 71 | |
NM_206885.2 | NP_996768.1 | 4 - 5 | 548 | 71 | |
NR_120441.1 | 3 - 4 | 468 | 71 | ||
NR_120442.1 | 3 - 4 | 468 | 71 | ||
NR_120443.1 | 3 - 4 | 468 | 71 | ||
NR_135801.1 | 4 - 5 | 548 | 71 | ||
NR_135802.1 | 4 - 5 | 548 | 71 | ||
XM_011516170.2 | XP_011514472.1 | 4 - 5 | 1338 | 71 | |
GenBank mRNA | AF523354.1 | - | 4 - 5 | 522 | 71 |
AY256823.1 | - | 4 - 5 | 522 | 71 | |
AY256824.1 | - | 4 - 5 | 522 | 71 | |
AY256825.1 | - | 4 - 5 | 522 | 71 | |
AY289133.1 | - | 2 - 3 | 294 | 71 | |
AY289134.1 | - | 2 - 3 | 294 | 71 | |
BC100832.1 | - | 3 - 4 | 453 | 71 | |
BC100833.1 | - | 3 - 4 | 453 | 71 | |
BC100834.1 | - | 3 - 4 | 453 | 71 | |
BC100835.1 | - | 3 - 4 | 453 | 71 |