Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244766.1 | NP_001231695.1 | 4 - 5 | 585 | 63 |
NM_001244767.1 | NP_001231696.1 | 3 - 4 | 473 | 63 | |
NM_173468.3 | NP_775739.1 | 3 - 4 | 473 | 63 | |
XM_005265709.2 | XP_005265766.1 | 5 - 6 | 654 | 63 | |
XM_011532412.2 | XP_011530714.1 | 3 - 4 | 474 | 63 | |
XM_017008837.1 | XP_016864326.1 | 3 - 4 | 384 | 63 | |
GenBank mRNA | AJ577473.1 | - | 2 - 3 | 276 | 63 |
AK093545.1 | - | 3 - 4 | 466 | 63 | |
AK299481.1 | - | 4 - 5 | 547 | 63 | |
AK313513.1 | - | 3 - 4 | 358 | 63 | |
BC038112.1 | - | 3 - 4 | 426 | 63 |