Product Details

Assay Reference Genome
Location

Chr.1:16987088 on build GRCh38
Cytoband
1p36.13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23400
Gene Symbol
ATP13A2
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Location
Chr.1:16985958-17011972 on build GRCh38
Assay Gene Location
Overlaps Intron 26 - Exon 26
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ATP13A2 NM_001141973.2 NP_001135445.1
NM_001141974.2 NP_001135446.1
NM_022089.3 NP_071372.1
XM_005245810.1 XP_005245867.1
XM_005245811.1 XP_005245868.1
XM_005245812.1 XP_005245869.1
XM_005245815.1 XP_005245872.1
XM_006710512.1 XP_006710575.1
XM_006710513.1 XP_006710576.1
XM_011541128.1 XP_011539430.1
XM_011541129.1 XP_011539431.1
XM_017000844.1 XP_016856333.1
XM_017000845.1 XP_016856334.1
XM_017000846.1 XP_016856335.1
XM_017000847.1 XP_016856336.1
XM_017000848.1 XP_016856337.1
XM_017000849.1 XP_016856338.1
XM_017000850.1 XP_016856339.1
AJ009947.1 CAA08912.1
AK025928.1
AK075310.1 BAC11539.1
AK290210.1
AL354615.1 CAB89728.1
AL833966.1 CAD38813.2
AY461712.1 AAR23423.1
BC030267.1 AAH30267.1
BC034575.1 AAH34575.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv508947 Chr1:16909817 - 17011115 on Build GRCh38 Insertion MFAP2 ATP13A2 CROCC
nsv954854 Chr1:16961106 - 17013905 on Build GRCh38 Deletion MFAP2 ATP13A2 CROCC
nsv428421 Chr1:16437838 - 17157486 on Build GRCh38 Gain+Loss FAM231B RNU1-1 PADI2 NBPF1 SDHB NECAP2 CROCCP2 RNU1-2 MST1P2 ATP13A2 CROCCP3 LOC105376805 SPATA21 RNU1-4 LOC105376796 MST1L RNU1-3 MIR3675 MFAP2 LOC102723383 ESPNP LOC100132147 CROCC FAM231A
dgv108n100 Chr1:16850538 - 17010370 on Build GRCh38 Gain MIR3675 MFAP2 RNU1-2 ATP13A2 CROCC LOC105376805
dgv92n100 Chr1:16684943 - 17003642 on Build GRCh38 Gain MIR3675 MFAP2 RNU1-2 LOC102723383 ATP13A2 ESPNP CROCC LOC105376805 RNU1-4 MST1L
nsv834280 Chr1:16888218 - 17123006 on Build GRCh38 Loss PADI2 SDHB MFAP2 RNU1-2 ATP13A2 CROCC LOC105376805
dgv120n100 Chr1:16890896 - 16997938 on Build GRCh38 Gain MFAP2 RNU1-2 ATP13A2 CROCC
nsv834269 Chr1:16855283 - 17038531 on Build GRCh38 Loss MIR3675 SDHB MFAP2 RNU1-2 ATP13A2 CROCC LOC105376805

View Full Product Details