Product Details

Assay Reference Genome
Location

Chr.16:88811872 on build GRCh38
Cytoband
16q24.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
353
Gene Symbol
APRT
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
AMP, APRTD
Location
Chr.16:88809469-88811934 on build GRCh38
Assay Gene Location
Overlaps Intron 1 - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
APRT NM_000485.2 NP_000476.1
NM_001030018.1 NP_001025189.1
BC107151.2
BM550173.1
CR749423.1 CAH18261.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n68 Chr16:88767460 - 88944903 on Build GRCh38 Loss LOC100129697 TRAPPC2L GALNS LOC105371409 PIEZO1 LOC107987238 CBFA2T3 LOC101927793 CDT1 APRT PABPN1L
nsv1063024 Chr16:88707330 - 89238482 on Build GRCh38 Gain GALNS ZNF778 LOC105371409 LOC100289580 ACSF3 SLC22A31 LOC101927793 LINC00304 CDH15 LOC100129697 CTU2 TRAPPC2L LOC400558 MIR4722 PIEZO1 LOC107987238 CBFA2T3 CDT1 LOC339059 APRT PABPN1L
nsv471115 Chr16:88801844 - 89197442 on Build GRCh38 Loss GALNS LOC105371409 ACSF3 SLC22A31 LOC101927793 LINC00304 CDH15 LOC100129697 TRAPPC2L LOC400558 LOC107987238 CBFA2T3 CDT1 APRT PABPN1L
dgv3064n100 Chr16:88683753 - 88846491 on Build GRCh38 Gain CTU2 GALNS MIR4722 PIEZO1 LOC107987238 RNF166 SNAI3-AS1 LOC100289580 CDT1 LOC339059 SNAI3 APRT
nsv573674 Chr16:88738775 - 89325907 on Build GRCh38 Gain GALNS ZNF778 LOC105371409 LOC100289580 ACSF3 SLC22A31 LOC101927793 LINC00304 CDH15 LOC100129697 TRAPPC2L LOC105371414 LOC400558 LOC100287036 PIEZO1 LOC107987238 ANKRD11 CBFA2T3 CDT1 LOC339059 APRT PABPN1L
esv3639555 Chr16:88745961 - 88899576 on Build GRCh38 Gain TRAPPC2L GALNS PIEZO1 LOC107987238 CBFA2T3 LOC101927793 CDT1 APRT PABPN1L
nsv471117 Chr16:88805821 - 88838392 on Build GRCh38 Gain GALNS LOC107987238 CDT1 APRT
nsv952071 Chr16:88790893 - 89019992 on Build GRCh38 Deletion LOC100129697 TRAPPC2L GALNS LOC105371409 LOC107987238 CBFA2T3 LOC101927793 CDT1 APRT PABPN1L
nsv482951 Chr16:88633593 - 90228345 on Build GRCh38 Loss LOC105371419 ZNF778 CENPBD1 CYBA SPATA2L LOC100129697 TUBB8P7 ZNF276 MIR4722 LOC100287036 CHMP1A URAHP SPATA33 AFG3L1P TUBB3 LOC339059 PABPN1L CPNE7 GALNS LOC107984817 RPL13 MC1R LOC105371409 DEF8 LOC100289580 GAS8 CDH15 TRAPPC2L LOC105371414 LOC400558 LOC107987238 FAM157C SNAI3 IL17C LOC101927817 DPEP1 FANCA VPS9D1 SPG7 ACSF3 LOC101927793 LINC00304 CTU2 PIEZO1 RNF166 SNAI3-AS1 ANKRD11 SPIRE2 DBNDD1 SNORD68 GAS8-AS1 SLC22A31 VPS9D1-AS1 PRDM7 CBFA2T3 TCF25 CDK10 CDT1 APRT MVD
nsv1063441 Chr16:88767066 - 88861882 on Build GRCh38 Loss TRAPPC2L GALNS PIEZO1 LOC107987238 CDT1 APRT
dgv3065n100 Chr16:88711587 - 88927172 on Build GRCh38 Gain GALNS LOC100289580 LOC101927793 CTU2 TRAPPC2L MIR4722 PIEZO1 LOC107987238 CBFA2T3 CDT1 LOC339059 APRT PABPN1L

View Full Product Details