Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016176.3 | NP_057260.2 | 4 - 5 | 66 | |
NM_016547.2 | NP_057631.1 | 4 - 5 | 66 | ||
GenBank mRNA | AF132749.1 | - | 4 - 5 | 824 | 66 |
AF178986.1 | - | 4 - 5 | 874 | 66 | |
AK027277.1 | - | 5 - 6 | 502 | 66 | |
AK075352.1 | - | 4 - 5 | 844 | 66 | |
AK223079.1 | - | 4 - 5 | 826 | 66 | |
BC006211.2 | - | 4 - 5 | 791 | 66 | |
BC007625.1 | - | 3 - 4 | 659 | 66 | |
BC008917.2 | - | 4 - 5 | 810 | 66 | |
BC011244.1 | - | 4 - 5 | 764 | 66 | |
BC022375.1 | - | 4 - 5 | 844 | 66 | |
L79912.1 | - | 3 - 4 | 589 | 66 |