Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270679.1 | NP_001257608.1 | 1 - 2 | 171 | 103 |
NM_138357.2 | NP_612366.1 | 1 - 2 | 171 | 103 | |
GenBank mRNA | AK075439.1 | - | 1 - 2 | 159 | 103 |
AK315519.1 | - | 1 - 2 | 154 | 103 | |
BC010682.1 | - | 1 - 2 | 128 | 103 | |
BC034235.1 | - | 1 - 2 | 171 | 103 | |
HY146050.1 | - | 1 - 2 | 173 | 103 |