Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007349.3 | NP_031375.3 | 5 - 6 | 480 | 84 |
XM_005249539.1 | XP_005249596.1 | 6 - 7 | 619 | 84 | |
XM_011515982.2 | XP_011514284.1 | 5 - 6 | 594 | 84 | |
XM_011515983.1 | XP_011514285.1 | 5 - 6 | 480 | 84 | |
XM_017011881.1 | XP_016867370.1 | 6 - 7 | 1792 | 84 | |
GenBank mRNA | AK123044.1 | - | 1 - 2 | 525 | 84 |
AK123600.1 | - | 6 - 7 | 619 | 84 | |
AK293744.1 | - | 3 - 4 | 312 | 84 | |
AK307417.1 | - | 5 - 6 | 483 | 84 | |
BQ222021.1 | - | - | 424 | 84 |