Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 10 - 11 | 1384 | 61 |
XM_005272599.3 | XP_005272656.1 | 10 - 11 | 1313 | 61 | |
GenBank mRNA | AJ005890.1 | - | 10 - 11 | 1300 | 61 |
AK298264.1 | - | 10 - 11 | 1379 | 61 | |
AK316375.1 | - | 10 - 11 | 1330 | 61 | |
BC000972.2 | - | 10 - 11 | 1331 | 61 | |
BC011675.2 | - | 10 - 11 | 1215 | 61 |