Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 16 - 17 | 1943 | 72 |
XM_005272599.3 | XP_005272656.1 | 16 - 17 | 1872 | 72 | |
GenBank mRNA | AJ005890.1 | - | 16 - 17 | 1859 | 72 |
AK291976.1 | - | 16 - 17 | 1904 | 72 | |
BC000972.2 | - | 16 - 17 | 1890 | 72 | |
BC011675.2 | - | 16 - 17 | 1774 | 72 |