Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 3 - 4 | 527 | 58 |
XM_005272599.3 | XP_005272656.1 | 3 - 4 | 459 | 58 | |
GenBank mRNA | AJ005890.1 | - | 3 - 4 | 443 | 58 |
AK291976.1 | - | 3 - 4 | 488 | 58 | |
AK296911.1 | - | 3 - 4 | 498 | 58 | |
AK298264.1 | - | 3 - 4 | 522 | 58 | |
AK316375.1 | - | 3 - 4 | 473 | 58 | |
BC000972.2 | - | 3 - 4 | 474 | 58 | |
BC011675.2 | - | 3 - 4 | 358 | 58 | |
BG912845.1 | - | 3 - 4 | 513 | 58 |