Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014008.4 | NP_054727.1 | 4 - 5 | 637 | 63 |
XM_005272599.3 | XP_005272656.1 | 4 - 5 | 569 | 63 | |
GenBank mRNA | AJ005890.1 | - | 4 - 5 | 553 | 63 |
AK291976.1 | - | 4 - 5 | 598 | 63 | |
AK296911.1 | - | 4 - 5 | 608 | 63 | |
AK298264.1 | - | 4 - 5 | 632 | 63 | |
AK316375.1 | - | 4 - 5 | 583 | 63 | |
BC000972.2 | - | 4 - 5 | 584 | 63 | |
BC011675.2 | - | 4 - 5 | 468 | 63 |