Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003725.3 | NP_003716.2 | 2 - 3 | 526 | 76 |
XM_005269207.1 | XP_005269264.1 | 3 - 4 | 675 | 76 | |
XM_005269208.1 | XP_005269265.1 | 3 - 4 | 679 | 76 | |
XM_005269209.1 | XP_005269266.1 | 3 - 4 | 666 | 76 | |
XM_006719672.1 | XP_006719735.1 | 3 - 4 | 674 | 76 | |
XM_011538925.1 | XP_011537227.1 | 3 - 4 | 669 | 76 | |
XM_011538926.1 | XP_011537228.1 | 3 - 4 | 659 | 76 | |
XM_011538927.1 | XP_011537229.1 | 3 - 4 | 650 | 76 | |
GenBank mRNA | AF016509.1 | - | 1 - 2 | 310 | 76 |
AF223225.1 | - | 2 - 3 | 397 | 76 | |
AK313423.1 | - | 2 - 3 | 436 | 76 | |
BC020710.1 | - | 2 - 3 | 420 | 76 |