Assay Design



Gene Transcripts

Gene Symbol
NOVA2
Entrez Gene ID
4858
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
ANOVA, NOVA3
Chromosome Location
Chr.19: 45933734 - 45973399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002516.3 NP_002507.1 1 - 2 137 55
GenBank mRNA AF083898.1 - 1 - 2 137 55
U70477.1 - 1 - 2 108 55

View Full Product Details