Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020810.3 | NP_065861.3 | 1 - 2 | 118 | 111 |
GenBank mRNA | AK293158.1 | - | 1 - 2 | 88 | 111 |
BC108284.1 | - | 1 - 2 | 89 | 111 | |
BQ050607.1 | - | 1 - 2 | 96 | 111 | |
DB448202.2 | - | 1 - 2 | 104 | 111 | |
DC370229.1 | - | 1 - 2 | 118 | 111 |