Assay Design



Gene Transcripts

Gene Symbol
TRMT5
Entrez Gene ID
57570
Gene Name
tRNA methyltransferase 5
Gene Aliases
COXPD26, KIAA1393, TRM5
Chromosome Location
Chr.14: 60971441 - 60981170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020810.3 NP_065861.3 4 - 5 1548 79
XM_005267916.3 XP_005267973.2 4 - 5 1578 79
XM_011537017.2 XP_011535319.1 4 - 5 1639 79
GenBank mRNA AB037814.1 - 3 - 4 1417 79
AK293158.1 - 4 - 5 1518 79
BC108284.1 - 4 - 5 1519 79
BC136606.1 - 4 - 5 1463 79
BC136607.1 - 4 - 5 1463 79

View Full Product Details