Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258017.1 | NP_001244946.1 | 5 - 6 | 859 | 85 |
NM_001258018.1 | NP_001244947.1 | 5 - 6 | 941 | 85 | |
NM_152899.1 | NP_690863.1 | 3 - 4 | 332 | 85 | |
NM_172374.2 | NP_758962.1 | 5 - 6 | 883 | 85 | |
NR_047577.1 | 6 - 7 | 918 | 85 | ||
GenBank mRNA | AF293462.1 | - | 3 - 4 | 332 | 85 |
AJ880386.1 | - | 5 - 6 | 758 | 85 | |
AJ880387.1 | - | 5 - 6 | 735 | 85 | |
AJ880388.1 | - | 5 - 6 | 797 | 85 | |
AJ880389.1 | - | 6 - 7 | 795 | 85 | |
AY358933.1 | - | 3 - 4 | 332 | 85 | |
BC026103.1 | - | 5 - 6 | 787 | 85 | |
BC064378.1 | - | 3 - 4 | 318 | 85 | |
BC090852.1 | - | 3 - 4 | 319 | 85 | |
BC131625.1 | - | 5 - 6 | 772 | 85 | |
DQ079587.1 | - | 5 - 6 | 883 | 85 | |
DQ079588.1 | - | 5 - 6 | 941 | 85 | |
DQ079589.1 | - | 5 - 6 | 859 | 85 |