Assay Design



Gene Transcripts

Gene Symbol
ZFP1
Entrez Gene ID
162239
Gene Name
ZFP1 zinc finger protein
Gene Aliases
ZNF475
Chromosome Location
Chr.16: 75126285 - 75172234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318469.1 NP_001305398.1 1 - 2 222 62
NM_001318473.1 NP_001305402.1 1 - 2 222 62
GenBank mRNA BC064604.1 - 1 - 2 81 62

View Full Product Details