Assay Design



Gene Transcripts

Gene Symbol
NAA40
Entrez Gene ID
79829
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
NAT11, PATT1
Chromosome Location
Chr.11: 63938970 - 63957322 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024771.3 NP_079047.2 1 - 2 127 66
XM_006718689.3 XP_006718752.1 1 - 2 114 66
XM_011545254.2 XP_011543556.1 1 - 2 124 66
XM_011545255.2 XP_011543557.1 1 - 2 124 66
GenBank mRNA AK300451.1 - 1 - 2 101 66
AK303557.1 - 1 - 2 66
BC041617.1 - 1 - 2 56 66
BC052298.1 - 1 - 2 56 66
BX647309.1 - 1 - 2 138 66

View Full Product Details