Assay Design



Gene Transcripts

Gene Symbol
RBM25
Entrez Gene ID
58517
Gene Name
RNA binding motif protein 25
Gene Aliases
NET52, RED120, RNPC7, S164, Snu71, fSAP94
Chromosome Location
Chr.14: 73058436 - 73121675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021239.2 NP_067062.1 14 - 15 1900 67
XM_011537044.2 XP_011535346.1 15 - 16 2002 67
XM_011537045.2 XP_011535347.1 14 - 15 1812 67
GenBank mRNA AK094697.1 - 13 - 14 1744 67
AL832314.1 - 13 - 14 5316 67
AL833497.1 - 2 - 3 749 67
BC113389.1 - 13 - 14 1680 67
BC113391.1 - 13 - 14 1680 67
BC136775.1 - 14 - 15 1737 67
BC136776.1 - 14 - 15 1737 67
BC144407.1 - 15 - 16 1805 67
BX647116.1 - 14 - 15 1853 67

View Full Product Details