Assay Design



Gene Transcripts

Gene Symbol
QPRT
Entrez Gene ID
23475
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
HEL-S-90n, QPRTase
Chromosome Location
Chr.16: 29670588 - 29698699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014298.4 NP_055113.2 1 - 2 199 125
NR_134536.1 1 - 2 199 125
XM_005255223.3 XP_005255280.2 2 - 3 282 125
XM_017023101.1 XP_016878590.1 2 - 3 282 125
GenBank mRNA BC005060.1 - 1 - 2 36 125
BC010033.2 - 1 - 2 28 125
BC018910.2 - 1 - 2 53 125
BG748194.1 - 1 - 2 170 124
BJ997057.1 - 1 - 2 62 125
EU794638.1 - 1 - 2 28 125

View Full Product Details