Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007056.2 | NP_008987.2 | 2 - 3 | 201 | 65 |
NR_103529.1 | 2 - 3 | 201 | 65 | ||
XM_011526396.2 | XP_011524698.1 | 2 - 3 | 196 | 65 | |
XM_011526397.2 | XP_011524699.1 | 1 - 2 | 46 | 65 | |
GenBank mRNA | AF042800.1 | - | 2 - 3 | 201 | 65 |
AK094681.1 | - | 1 - 2 | 795 | 65 | |
AY358944.1 | - | 2 - 3 | 182 | 65 | |
BC013178.1 | - | 2 - 3 | 146 | 65 | |
BC080554.1 | - | 1 - 2 | 141 | 65 | |
BC131496.1 | - | 1 - 2 | 59 | 65 |