Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159944.2 | NP_001153416.1 | 9 - 10 | 1191 | 61 |
NM_145245.4 | NP_660288.1 | 10 - 11 | 1302 | 61 | |
XM_005272458.4 | XP_005272515.1 | 10 - 11 | 1296 | 61 | |
XM_017026239.1 | XP_016881728.1 | 10 - 11 | 1288 | 61 | |
GenBank mRNA | AB449874.1 | - | 9 - 10 | 1148 | 61 |
AK091609.1 | - | 4 - 5 | 340 | 61 | |
BC014111.2 | - | 10 - 11 | 1220 | 61 |