Assay Design



Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
Chromosome Location
Chr.2: 64524305 - 64593005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002243.2 NP_001002243.1 2 - 3 2269 67
NM_017657.4 NP_060127.3 2 - 3 2269 67
NM_203437.3 NP_982261.2 2 - 3 2269 67
XM_005264380.1 XP_005264437.1 2 - 3 2269 67
GenBank mRNA AK000087.1 - 2 - 3 2244 67
AK021899.1 - 1 - 2 892 67
AK222821.1 - 2 - 3 2192 67
AL833962.2 - 2 - 3 2047 67
AY367088.1 - 1 - 2 1929 67
BC047529.1 - 2 - 3 2243 67

View Full Product Details