Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007191.4 | NP_009122.2 | 7 - 8 | 1205 | 59 |
GenBank mRNA | AF122922.1 | - | 7 - 8 | 942 | 59 |
AK301818.1 | - | 7 - 8 | 1132 | 59 | |
AY358344.1 | - | 7 - 8 | 975 | 59 | |
BC018037.1 | - | 7 - 8 | 948 | 59 | |
BX647427.1 | - | 7 - 8 | 874 | 59 |