Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007191.4 | NP_009122.2 | 9 - 10 | 1399 | 64 |
GenBank mRNA | AF122922.1 | - | 9 - 10 | 1136 | 64 |
AK301818.1 | - | 9 - 10 | 1326 | 64 | |
AY358344.1 | - | 9 - 10 | 1169 | 64 | |
BC018037.1 | - | 9 - 10 | 1142 | 64 | |
BX647427.1 | - | 9 - 10 | 1068 | 64 |