Assay Design
Gene Transcripts
- Gene Symbol
- RTEL1
- Entrez Gene ID
-
51750
- Gene Name
- regulator of telomere elongation helicase 1
- Gene Aliases
-
C20orf41, DKCA4, DKCB5, NHL, PFBMFT3, RTEL
- Chromosome Location
-
Chr.20: 63657810 - 63696253 on Build GRCh38
- Gene Symbol
- RTEL1-TNFRSF6B
- Entrez Gene ID
-
100533107
- Gene Name
- RTEL1-TNFRSF6B readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 63657810 - 63698698 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037882.1 |
|
21 - 22 |
2626 |
99 |
View Full Product Details