Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002440.3 | NP_002431.2 | 18 - 19 | 2645 | 85 |
GenBank mRNA | AF104243.1 | - | 18 - 19 | 2541 | 85 |
AJ844635.1 | - | 1 - 2 | 118 | 85 | |
BC033030.1 | - | 18 - 19 | 2635 | 85 | |
U89293.1 | - | 18 - 19 | 2571 | 85 |