Assay Design



Gene Transcripts

Gene Symbol
MSH4
Entrez Gene ID
4438
Gene Name
mutS homolog 4
Gene Aliases
-
Chromosome Location
Chr.1: 75796871 - 75913238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002440.3 NP_002431.2 4 - 5 814 131
GenBank mRNA AF104243.1 - 4 - 5 710 131
BC033030.1 - 4 - 5 804 131
U89293.1 - 4 - 5 740 131

View Full Product Details