Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002440.3 | NP_002431.2 | 9 - 10 | 1426 | 90 |
GenBank mRNA | AF104243.1 | - | 9 - 10 | 1322 | 90 |
BC033030.1 | - | 9 - 10 | 1416 | 90 | |
U89293.1 | - | 9 - 10 | 1352 | 90 |