Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077491.1 | NP_001070959.1 | 6 - 7 | 948 | 72 |
NM_001077492.1 | NP_001070960.1 | 5 - 6 | 880 | 72 | |
NM_012427.4 | NP_036559.1 | 5 - 6 | 1083 | 72 | |
XM_011526702.1 | XP_011525004.1 | 5 - 6 | 822 | 72 | |
XM_011526703.2 | XP_011525005.1 | 6 - 7 | 902 | 72 | |
GenBank mRNA | AF168768.2 | - | 5 - 6 | 899 | 72 |
AF435980.1 | - | 5 - 6 | 1054 | 72 | |
AF435981.1 | - | 5 - 6 | 851 | 72 | |
AK223421.1 | - | 5 - 6 | 1031 | 72 | |
AY279380.1 | - | 5 - 6 | 857 | 72 | |
AY279381.1 | - | 6 - 7 | 925 | 72 | |
AY359010.1 | - | 5 - 6 | 1083 | 72 | |
AY461805.1 | - | 3 - 4 | 826 | 72 | |
BC008036.2 | - | 5 - 6 | 1014 | 72 | |
BT006867.1 | - | 4 - 5 | 730 | 72 |