Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289177.1 | NP_001276106.1 | 12 - 13 | 1992 | 98 |
NM_001289178.1 | NP_001276107.1 | 12 - 13 | 1938 | 98 | |
NM_001289180.1 | NP_001276109.1 | 12 - 13 | 1702 | 98 | |
NM_001321384.1 | NP_001308313.1 | 12 - 13 | 1973 | 98 | |
NM_030924.4 | NP_112186.3 | 12 - 13 | 1983 | 98 | |
XM_011528329.1 | XP_011526631.1 | 9 - 10 | 1409 | 98 | |
XM_017027332.1 | XP_016882821.1 | 12 - 13 | 1810 | 98 | |
XM_017027333.1 | XP_016882822.1 | 12 - 13 | 1965 | 98 | |
XM_017027334.1 | XP_016882823.1 | 11 - 12 | 1797 | 98 | |
GenBank mRNA | AJ577571.1 | - | 12 - 13 | 1886 | 98 |
AK057412.1 | - | 7 - 8 | 1194 | 98 | |
AK093433.1 | - | 12 - 13 | 1951 | 98 | |
AK225862.1 | - | 11 - 12 | 1691 | 98 | |
AL136854.1 | - | 12 - 13 | 1724 | 98 | |
AY009107.1 | - | 12 - 13 | 1695 | 98 | |
AY358766.1 | - | 12 - 13 | 1983 | 98 | |
BC022027.1 | - | 12 - 13 | 1914 | 98 | |
BC090046.1 | - | 12 - 13 | 1899 | 98 |