Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198868.1 | NP_001185797.1 | 6 - 7 | 949 | 119 |
NM_001198869.1 | NP_001185798.1 | 6 - 7 | 918 | 119 | |
NM_005186.3 | NP_005177.2 | 6 - 7 | 934 | 119 | |
NR_040008.1 | 5 - 6 | 864 | 119 | ||
XM_006718698.1 | XP_006718761.1 | 6 - 7 | 849 | 119 | |
XM_011545292.1 | XP_011543594.1 | 6 - 7 | 860 | 119 | |
GenBank mRNA | AK022319.1 | - | - | 402 | 119 |
AK301538.1 | - | 5 - 6 | 756 | 119 | |
AK311001.1 | - | 5 - 6 | 504 | 119 | |
AK315556.1 | - | 6 - 7 | 863 | 119 | |
AY550975.1 | - | 5 - 6 | 757 | 119 | |
BC008751.2 | - | 6 - 7 | 885 | 119 | |
BC017200.2 | - | 6 - 7 | 771 | 119 | |
BC075862.1 | - | 6 - 7 | 852 | 119 | |
X04366.1 | - | 6 - 7 | 895 | 119 |