Assay Design



Gene Transcripts

Gene Symbol
CLIP3
Entrez Gene ID
25999
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
CLIPR-59, CLIPR59, RSNL1
Chromosome Location
Chr.19: 36014660 - 36032895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199570.1 NP_001186499.1 10 - 11 1765 101
NM_015526.2 NP_056341.1 11 - 12 1681 101
GenBank mRNA AJ427922.1 - 11 - 12 1659 101
AK056445.1 - 9 - 10 1164 101
AK090732.1 - 11 - 12 1684 101
AK092354.1 - 7 - 8 928 101
AK094738.1 - 11 - 12 1679 101
AK289509.1 - 10 - 11 1733 101
AK294748.1 - 11 - 12 1552 101
AL117468.1 - 3 - 4 293 101
BC013116.1 - 11 - 12 1593 101
BC014486.1 - 8 - 9 1124 101

View Full Product Details